Phosphate Buffered Saline (PBS), 10X, SeraCare
Supplier: MetaSystems Hard&Software GmbH
5460-0022 5460-0023
95059-252EA 179.32 USD
95059-252 95059-254
Phosphate Buffered Saline (PBS), 10X, SeraCare
Buffers Assay Buffers
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications.
  • Conductivity controlled