Human Recombinant IL-27B (from E. coli)
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-B66 009-001-B66-0005
RL009-001-B66EA 352.09 USD InStock
RL009-001-B66 10801-230 10802-560 10802-562
Human Recombinant IL-27B (from E. coli)
Proteins and Peptides
Epstein-Barr Virus Induced Gene-3 (EBI-3), is a secreted glycoprotein belonging to the hematopoietin receptor family and related to the p40 subunit of IL-12. It was identified by its induced expression in B-lymphocytes in response to Epstein-Barr virus infection. EBI-3 forms heterodimers with p28 to form IL-27 and with p35 to form IL-35. Both IL-27 and IL-35 have anti-inflammatory and regulatory activity. Recombinant Human EBI is a non-glycosylated polypeptide chain consisting of 209 amino acids with a molecular weight of 23.3 kDa.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty