Human Recombinant GDF15 (from E. coli)
Supplier: Bandelin Electronic GmbH &Co.KG
009-001-V09-0100 009-001-V09-0020 009-001-V09-0005
10801-560EA 6248 USD InStock
10801-560 10801-562 10801-564 10801-566
Human Recombinant GDF15 (from E. coli)
Proteins and Peptides
Growth and Differentiation Factor 15 (GDF-15) is a TGFβ family member, made by the placenta and heart tissues, that has a role in regulating inflammatory and apoptotic pathways. GDF-15 has become an immerging marker of early heart disease and has the potential as being used as a molecule for screening patients for early heart failure. Recombinant human GDF-15 D is a non-glycosylated, disulfide linked homodimer, containing two identical 113 amino acid chains, with a total molecular weight of 24.5 kDa. There is a His to an Asp substitution at position 7.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty