Maltose Binding Protein (MBP) Control Protein
Supplier: Bandelin Electronic GmbH &Co.KG
000-001-385-1 000-001-385-5
75929-874EA 1020.75 USD InStock
75929-874 75929-876
Maltose Binding Protein (MBP) Control Protein
Proteins and Peptides
Maltose Binding Protein (MBP) Control Protein
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Maltose-Binding Protein (MBP) is a part of the maltose/maltodextrin system of Escherichia coli, which is responsible for the uptake and efficient catabolism of maltodextrins. It is a complex regulatory and transport system involving many proteins and protein complexes. MBP has an approximate molecular mass of 42.5 kilodaltons. MBP is used to increase the solubility of recombinant proteins expressed in E. coli. In these systems, the protein of interest is often expressed as a MBP-fusion protein, preventing aggregation of the protein of interest.
Maltose-Binding Protein is suitable for use as a control in many assays including affinity purification, western blot and other biological assays. Optimal concentration should be determined by the researcher.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty