Mouse IL-13
Supplier: Bandelin Electronic GmbH &Co.KG
010-001-V52-0010 010-001-V52-0100 010-001-V52-0002
75929-056EA 8070 USD
75929-056 75934-668 75934-672 75934-676
Mouse IL-13
Proteins and Peptides
Recombinant Mouse IL-13 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-13 (IL-13) is an important cytokine secreted from Th2 cells. The functions attributed to IL-13 overlap significantly with those of IL-4 (induces IgE secretion from B cells and inhibits expression of inflammatory cytokines such as IL-1β, TNF-α, IL-8 and IL-6), but differs from IL-4 in that IL-13 seems to link inflammatory response of immune cells to the pathophysiological changes in the surrounding non-immune cells. The receptor subunits of IL-13 consist of IL-4Rα, IL-13Rα1 and IL-13Rα2. Human and mouse IL-13 are cross-reactive. Recombinant mouse IL-13 is a non-glycosylated protein, containing 111 amino acids, with a molecular weight of 12.3 kDa.
Interleukin-13 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-13 in immunological assays.