Oil Grip Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
NF35/7S NF35/8M NF35/9L NF35/10XL NF35/11XXL
10146-128CS 76.08 USD
10146-128 10146-130 10146-132 10146-134 10146-136
Oil Grip Gloves, Honeywell Safety
Gloves
For the handling of fine products in a greasy or damp environment.
  • Ergonomic shape
  • Washable
  • Superior, liquidproof grip
COMFORT. The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength, durability, superior comfort and ventilation, while enabling longer use and less fatigue.
RESISTANCE. The Polyamide yarn reduces linting for the most sensitive applications and can be easily laundered for extended use. The microfinish nitrile coating provides superior grip thanks to thousands of microscopic pockets that create a vacuum that disperses fluids and allows the entire grip surface to make clean contact with the object. It stays flexible and liquid proof in contact with oil.
HYGIENE. The dark colors make the glove less dirty. The open back provides superior ventilation.
Certificaciones: EN 388 - 3121 mechanical hazards. EN 0075/797/162/02/13/0158.