Nuts and Ferrules for PFA Tubing, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0777283 N0777284 N0777297 N8145237 N8145238
10139-314EA 248.07 USD
10139-314 10139-316 10139-338 10139-624 10139-626
Nuts and Ferrules for PFA Tubing, PerkinElmer
Fittings and Connectors
Ultra-clean fittings and PFA tubing are ideal for all trace metal analysis applications.
  • Easy to use
  • Ferrules install quickly, for high-flow applications
Low-flow valve fittings have low dead-volume, leak-free integrated ferrule connections.