Low-Profile Glove Dispensers, S-Curve
Supplier: S-Curve Technologies
AT-105 AT-105 PETG
10027-756EA 485.05 USD
10027-756 10027-870
Low-Profile Glove Dispensers, S-Curve
Glove Dispensers and Glove Box Holders
Low profile wall mount glove dispenser with front access holes. Designed to hold multiple glove sizes in a low-profile dispenser.
  • Sloped top to maintain HEPA airflow
  • Full-length hinge for durability
  • Mounts to wall
  • Wall bracket included
  • Flame treated edges for a smooth finish