International Labels 'Explosive Class 1', Labelmaster
Supplier: Label Master
SLEX33 SLEX32
101377-454PK 155.71 USD
101377-454 101377-452
International Labels 'Explosive Class 1', Labelmaster
Labels
Help meet the design and durability standards of 49 CFR, ICAO, and other international air regulations with vinyl International Wordless Explosive Class 1 labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives formulated for use in various climates and environments
  • Will not fade or fall off
Featuring a tab that includes all the compatibility letters to meet diverse needs, these labels help users stay in compliance with standards set forth by the DOT. Constructed of rugged, flexible material, they feature strong adhesion to painted, steel, fiber, and plastic surfaces. They will pass the 90-day IMO seawater immersion test.