Scissors, Dressing, Blunt Tipped Safety Blades, Excelta Corp®
Supplier: Excelta
278 376
75880-572EA 4.73 USD InStock
75880-572 75880-902 75880-974
Scissors, Dressing, Blunt Tipped Safety Blades, Excelta Corp®
Scissors
These scissors feature blunt tipped blades for safety.
  • Made of cleanroom safe surgical stainless steel (AISI 440)
They are ideal for dressing/dissection, and handling delicate material.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty