Sign, 'NOTICE PLEASE WASH HANDS BEFORE LEAVING' Accuform®
Supplier: ACCUFORM MANUFACTURING, INC
MRST815VS MRST815VA MRST815VP MHSK817VS MHSK817VA MHSK817VP
76442-474EA 16.05 USD
76442-474 76442-476 76442-478 76442-480 76442-482 76442-484
Sign, 'NOTICE PLEASE WASH HANDS BEFORE LEAVING' Accuform®
Signs
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage.
  • Multiple material and size options
Choose from three material options. Adhesive Vinyl signs feature a printed matte surface with permanent acrylic adhesive back. Moisture-resistant for a variety of light-duty indoor and sheltered outdoor marking needs. Plastic signs are durable with slight flex. Good material choice for indoor and sheltered outdoor environments. The printed surface has a clear UV protective overcoat. Aluminum signs provide the strength of metal without rusting. Printed surface protected by UV coating to withstand splashes, wash-downs, and outdoor conditions, plus abrasion and chemical resistance.