Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone 98%

Supplier: AMBEED, INC
Warning

A131152-1G A131152-100G A131152-25G A131152-5G A131152-1G
103801-532EA 39 USD InStock
103801-532 76870-748 76870-750 76870-752 77314-008
(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone 98%
(2-Butyl-5-nitrobenzofuran-3-yl)(4-methoxyphenyl)methanone
Precaución: for Research Use Only


Formula: C₂₀H₁₉NO₅
Boiling Pt: 537.726 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD14525623
CAS Number: 141627-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty