Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-methoxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A121579-1G A121579-10G A121579-250MG A121579-25G A121579-1G A121579-5G
103800-086EA 23 USD InStock
103800-086 77009-448 77009-450 77009-452 77312-208 77312-210
2-Chloro-3-methoxyphenylboronic acid 97%
2-Chloro-3-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD12913933
CAS Number: 854778-30-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty