Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,4,6-Tetra-O-acetyl-β-D-glucosamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A169339-1G A169339-5G A169339-10G A169339-250MG A169339-1G A169339-5G
103806-376EA 8 USD InStock
103806-376 103844-630 76813-164 76813-166 77300-972 77300-974
1,3,4,6-Tetra-O-acetyl-β-D-glucosamine hydrochloride 97%
1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₂ClNO₉
Melting Pt: 197…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075204
CAS Number: 10034-20-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty