Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone 97%

Supplier: AMBEED, INC
Warning

A121582-1G A121582-100G A121582-10G A121582-250MG A121582-25G A121582-1G A121582-5G
103800-088EA 7 USD InStock
103800-088 77246-126 77246-128 77246-130 77246-132 77312-212 77312-214
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone 97%
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone
Precaución: for Research Use Only


Formula: C₈H₁₂O₅
Boiling Pt: 338.1±37.0 °C (760 mm Hg)
Melting Pt: 135…139 °C
Flash Pt: 139.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00080793
CAS Number: 30725-00-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty