Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Methylpyridin-2-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A131439-1G A131439-250MG A131439-100MG
103801-580EA 164 USD InStock
103801-580 76688-642 77624-116
(4-Methylpyridin-2-yl)methanol 97%
(4-Methylpyridin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₇H₉NO
Boiling Pt: 236 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD07774103
CAS Number: 42508-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty