Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-1-Phenyl-1,2-ethanediol 98%

Supplier: AMBEED, INC
Warning

A171802-1G A171802-5G A171802-10G A171802-1G A171802-25G A171802-5G
103806-666EA 9 USD InStock
103806-666 103844-944 76769-254 77082-114 77082-116 77082-118
(R)-(-)-1-Phenyl-1,2-ethanediol 98%
(R)-(-)-1-Phenyl-1,2-ethanediol
Precaución: for Research Use Only


Formula: C₈H₁₀O₂
Boiling Pt: 273.5 °C (760 mm Hg)
Melting Pt: 65…68 °C
Flash Pt: 145.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064262
CAS Number: 16355-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty