Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-cysteic acid 97%

Supplier: AMBEED, INC
Warning

A177979-1G A177979-1G A177979-5G A177979-250MG
103807-286EA 24 USD InStock
103807-286 103845-626 77083-022 77083-024 77191-608
Fmoc-L-cysteic acid 97%
Fmoc-L-cysteic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₇NO₇S
Storage Temperature: Refrigerator
MDL Number: MFCD02682540
CAS Number: 751470-47-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty