Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Methoxypropan-1-ol 95%

Supplier: AMBEED, INC
Danger

A856265-100MG A856265-250MG
103799-732EA 152 USD InStock
103799-732 76791-892 76791-894
(R)-2-Methoxypropan-1-ol 95%
(R)-2-Methoxypropan-1-ol
Precaución: for Research Use Only


Formula: C₄H₁₀O₂
Boiling Pt: 130.0±0.0 °C (760 mm Hg)
Flash Pt: 41.9 °C
Storage Temperature: Freezer
MDL Number: MFCD01632586
CAS Number: 6131-59-5
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty