Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A117080-1G A117080-100G A117080-10G A117080-25G A117080-1G A117080-5G
103798-824EA 5 USD InStock
103798-824 103824-696 76971-270 76971-272 76971-274 77311-482 77311-484
Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate 97%
Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
Precaución: for Research Use Only


Formula: C₆H₉NO₃
MW: 143.1406 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318621
CAS Number: 64700-65-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty