Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-3-Hydroxytetrahydrofuran 98%

Supplier: AMBEED, INC
Warning

A267309-1G A267309-5G A267309-1G A267309-25G A267309-5G A267309-100G A267309-10G A267309-500G
103812-844EA 9 USD InStock
103812-844 103852-744 77092-472 77092-474 77092-476 77140-916 77140-918 77140-920
(R)-(-)-3-Hydroxytetrahydrofuran 98%
(R)-(-)-3-Hydroxytetrahydrofuran
Precaución: for Research Use Only


Formula: C₄H₈O₂
Boiling Pt: 181 °C (760 mm Hg)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00067101
CAS Number: 86087-24-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty