Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thien-3-yl-L-alanine 97%

Supplier: AMBEED, INC
Warning

A244475-1G A244475-1G A244475-250MG A244475-5G
103811-584EA 151 USD InStock
103811-584 77090-626 77193-464 77193-466
3-Thien-3-yl-L-alanine 97%
3-Thien-3-yl-L-alanine
Precaución: for Research Use Only


Formula: C₇H₉NO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00038645
CAS Number: 3685-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty