Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Hydroxy-4-phenylbutyric acid 97%

Supplier: AMBEED, INC
Warning

A124038-1G A124038-10G A124038-250MG A124038-25G A124038-1G
103800-476EA 36 USD InStock
103800-476 76688-244 76688-246 76688-248 77312-604
(S)-2-Hydroxy-4-phenylbutyric acid 97%
(S)-2-Hydroxy-4-phenylbutyric acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.2005 g/mol
Boiling Pt: 356.9 °C (760 mm Hg)
Melting Pt: 110…112 °C
Flash Pt: 183.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06656492
CAS Number: 115016-95-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty