Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A175247-1G A175247-10G A175247-250MG A175247-1G A175247-25G A175247-5G
103807-014EA 23 USD InStock
103807-014 103845-318 76690-318 76690-320 77082-652 77082-654 77082-656
(S)-4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%
(S)-4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₈N₂O₄
Boiling Pt: 371.8 °C (760 mm Hg)
Melting Pt: 231…239 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179098
CAS Number: 848482-93-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty