Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Aminopyrimidin-4-yl)ethanone 95%

Supplier: AMBEED, INC
Warning

A355943-1G A355943-5G A355943-100MG A355943-250MG
103826-154EA 72 USD InStock
103826-154 103837-162 77015-768 77015-770
1-(2-Aminopyrimidin-4-yl)ethanone 95%
1-(2-Aminopyrimidin-4-yl)ethanone
Precaución: for Research Use Only


Formula: C₆H₇N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD09263814
CAS Number: 106157-82-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty