Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol 97%

Supplier: AMBEED, INC
Warning

A256178-5G A256178-5G A256178-100G
103852-106EA 12 USD InStock
103852-106 77289-136 77626-342
1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol 97%
1-(3,3-Diphenyl-N-methylpropylamino)-2-methyl-2-propanol
Precaución: for Research Use Only


Formula: C₂₀H₂₇NO
Boiling Pt: 428.9 °C (760 mm Hg)
Flash Pt: 164.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07782118
CAS Number: 100442-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty