Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dimethyl-1H-imidazole-4-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A310605-1G A310605-100MG A310605-250MG
103814-626EA 419 USD InStock
103814-626 103835-810 77881-864 77881-865
1,5-Dimethyl-1H-imidazole-4-carboxylic acid 97%
1,5-Dimethyl-1H-imidazole-4-carboxylic acid
Precaución: Sealed in dry, at 2…8°C


Formula: C₆H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18910969
CAS Number: 80304-42-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty