Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dimethyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A164440-1G A164440-5G A164440-250MG A164440-1G
103805-862EA 20 USD InStock
103805-862 103844-120 77191-204 77320-284
1,5-Dimethyl-1H-pyrazole-4-carbaldehyde 98%
1,5-Dimethyl-1H-pyrazole-4-carbaldehyde
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₆H₈N₂O
Boiling Pt: 229 °C (760 mm Hg)
Flash Pt: 92.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159630
CAS Number: 25711-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty