Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecanedioic acid monomethyl ester 97%

Supplier: AMBEED, INC
Warning

A202304-1G A202304-5G A202304-25G A202304-5G A202304-10G A202304-500G A202304-1G
103817-370EA 7 USD InStock
103817-370 103848-008 77086-114 77086-116 77097-416 77097-418 77302-802
Dodecanedioic acid monomethyl ester 97%
Dodecanedioic acid monomethyl ester
Precaución: for Research Use Only


Formula: C₁₃H₂₄O₄
Boiling Pt: 355.3 °C (760 mm Hg)
Flash Pt: 124.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00053323
CAS Number: 3903-40-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty