Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzylindole-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A935693-5G A935693-1G A935693-100MG A935693-10G A935693-250MG
103841-418EA 240 USD InStock
103841-418 76845-682 77281-990 77281-992 77281-994
1-Benzylindole-3-carboxylic acid 95%
1-Benzylindole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₃NO₂
Boiling Pt: 496.3 °C (760 mm Hg)
Flash Pt: 254 °C
Storage Temperature: Freezer
MDL Number: MFCD00057094
CAS Number: 27018-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty