Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-chloro-5-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A179832-1G A179832-5G A179832-1G A179832-25G A179832-10G A179832-250MG
103807-478EA 7 USD InStock
103807-478 103845-830 77083-302 77083-304 77236-532 77236-534
1-Bromo-3-chloro-5-iodobenzene 98%
1-Bromo-3-chloro-5-iodobenzene
Precaución: For research use only.


Formula: C₆H₃BrClI
Boiling Pt: 280.4 °C (760 mm Hg)
Melting Pt: 85.5 °C
Flash Pt: 123.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07778998
CAS Number: 13101-40-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty