Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(trans-4-propylcyclohexyl)benzene 96%

Supplier: AMBEED, INC
Warning

A213239-1G A213239-5G A213239-1G A213239-100G A213239-10G
103818-274EA 5 USD InStock
103818-274 103848-980 77087-476 77275-240 77275-242
1-Bromo-4-(trans-4-propylcyclohexyl)benzene 96%
1-Bromo-4-(trans-4-propylcyclohexyl)benzene
Precaución: for Research Use Only


Formula: C₁₅H₂₁Br
Boiling Pt: 332.1 °C (760 mm Hg)
Flash Pt: 149.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00577367
CAS Number: 86579-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty