Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexyl methyl ketone 97%

Supplier: AMBEED, INC
Warning

A279794-1G A279794-5G A279794-10G A279794-500G A279794-1G A279794-25G A279794-5G A279794-100G
103813-482EA 8 USD InStock
103813-482 103853-428 76692-996 76692-998 77305-342 77305-344 77305-346 77879-525
Cyclohexyl methyl ketone 97%
Cyclohexyl methyl ketone
Precaución: for Research Use Only


Formula: C₈H₁₄O
Storage Temperature: Refrigerator
MDL Number: MFCD00040418
CAS Number: 823-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty