Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A149888-1G A149888-5G A149888-100MG A149888-250MG A149888-1G A149888-5G
103804-096EA 20 USD InStock
103804-096 103842-222 77162-736 77162-738 77317-734 77317-736
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid 97%
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272240
CAS Number: 754214-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty