Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-N-Boc-3-(4-Bromophenoxy)pyrrolidine 97%

Supplier: AMBEED, INC
Warning

A128784-1G A128784-100MG A128784-250MG A128784-1G A128784-5G
103801-182EA 63 USD InStock
103801-182 77149-508 77149-510 77313-532 77313-534
1-N-Boc-3-(4-Bromophenoxy)pyrrolidine 97%
1-N-Boc-3-(4-Bromophenoxy)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₅H₂₀BrNO₃
Boiling Pt: 403.6±35.0 °C (760 mm Hg)
Flash Pt: 197.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12068410
CAS Number: 1159825-42-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty