Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,2,2-Trifluoroethoxy)phenol 97%

Supplier: AMBEED, INC
Warning

A300255-1G A300255-5G A300255-100G A300255-10G A300255-250MG A300255-1G A300255-5G
103814-266EA 16 USD InStock
103814-266 103835-418 76827-846 76827-848 76827-850 77305-800 77305-802
2-(2,2,2-Trifluoroethoxy)phenol 97%
2-(2,2,2-Trifluoroethoxy)phenol
Precaución: for Research Use Only


Formula: C₈H₇F₃O₂
Boiling Pt: 202 °C (760 mm Hg)
Melting Pt: 49…50 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD11181824
CAS Number: 160968-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty