Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA) 98%

Supplier: AMBEED, INC
Warning

A140665-5MG A140665-10MG A140665-50MG A140665-1G A140665-1MG A140665-10MG A140665-250MG
103791-476EA 8 USD InStock
103791-476 103792-172 103799-370 103802-892 76798-242 77315-998 77316-000 77566-832
2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA) 98%
2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA)
Precaución: For research use only.


Formula: C₁₇H₁₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00870555
CAS Number: 117570-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty