Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzo[b]thiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A142191-1G A142191-100MG A142191-10G A142191-250MG A142191-1G A142191-5G
103803-116EA 98 USD InStock
103803-116 77235-022 77235-024 77235-026 77316-332 77316-334
2-(Benzo[b]thiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(Benzo[b]thiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₄H₁₇BO₂S
Boiling Pt: 377.7 °C (760 mm Hg)
Melting Pt: 118 °C
Flash Pt: 182.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664405
CAS Number: 501945-71-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty