Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Hydroxyethyl)benzotrifluoride 97%

Supplier: AMBEED, INC
Warning

A589404-1G A589404-25G A589404-5G
103828-920EA 15 USD InStock
103828-920 103840-064 76844-208 76844-210 76844-212
2-(2-Hydroxyethyl)benzotrifluoride 97%
2-(2-Hydroxyethyl)benzotrifluoride
Precaución: for Research Use Only


Formula: C₉H₉F₃O
Boiling Pt: 185 °C (760 mm Hg)
Melting Pt: 220…223 °C at N/A mmHg
Flash Pt: 102.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00003540
CAS Number: 94022-96-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty