Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Bibenzo[d][1,3,2]dioxaborole 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A253241-1G A253241-10G A253241-250MG A253241-1G A253241-25G A253241-5G A253241-100G A253241-500G
103812-086EA 7 USD InStock
103812-086 103851-934 76801-590 76801-592 77091-360 77091-362 77091-364 77550-608 77668-066
2,2'-Bibenzo[d][1,3,2]dioxaborole 98% (by HPLC)
2,2'-Bibenzo[d][1,3,2]dioxaborole
Precaución: for Research Use Only


Formula: C₁₂H₈B₂O₄
Boiling Pt: 315 °C (760 mm Hg)
Melting Pt: 189…196 °C
Flash Pt: 144.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842343
CAS Number: 13826-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty