Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5-Trimethylcyclohexa-2,5-diene-1,4-dione 97%

Supplier: AMBEED, INC
Danger

A200164-1G A200164-5G A200164-25G A200164-1G
103817-170EA 5 USD InStock
103817-170 103847-798 77085-856 77302-656
2,3,5-Trimethylcyclohexa-2,5-diene-1,4-dione 97%
2,3,5-Trimethylcyclohexa-2,5-diene-1,4-dione
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
Boiling Pt: 215.3 °C (760 mm Hg)
Melting Pt: 31…33 °C
Flash Pt: 77.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00045537
CAS Number: 935-92-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty