Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,6,7,10,11-Hexamethoxytriphenylene 97%

Supplier: AMBEED, INC
Warning

A205066-1G A205066-5G A205066-10G A205066-250MG
103817-588EA 15 USD InStock
103817-588 103848-252 76873-542 76873-544
2,3,6,7,10,11-Hexamethoxytriphenylene 97%
2,3,6,7,10,11-Hexamethoxytriphenylene
Precaución: for Research Use Only


Formula: C₂₄H₂₄O₆
Boiling Pt: 578.6±45.0 °C (760 mm Hg)
Melting Pt: 300 °C
Flash Pt: 235.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00075571
CAS Number: 808-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty