Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A256278-1G A256278-5G A256278-10G A256278-250MG A256278-1G A256278-25G A256278-5G
103812-258EA 15 USD InStock
103812-258 103852-122 76786-884 76786-886 77091-608 77091-610 77091-612
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride 97%
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride
Precaución: for Research Use Only


Formula: C₇H₈N₂·2ClH
Flash Pt: 126.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15144657
CAS Number: 6000-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty