Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A260416-1G A260416-5G A260416-100MG A260416-10G A260416-250MG A260416-1G A260416-5G
103812-496EA 33 USD InStock
103812-496 103852-378 77113-666 77113-668 77113-670 77304-794 77304-796
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile 98%
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₇NO
Boiling Pt: 325.6 °C (760 mm Hg)
Melting Pt: 131.5…132.8 °C
Flash Pt: 150.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08234685
CAS Number: 25724-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty