Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dihydroxyterephthalic acid 97%

Supplier: AMBEED, INC
Warning

A644434-5G A644434-1G A644434-5G A644434-250MG A644434-100G
103840-364EA 42 USD InStock
103840-364 76844-534 76844-536 77035-416 77551-954
2,3-Dihydroxyterephthalic acid 97%
2,3-Dihydroxyterephthalic acid
Precaución: for Research Use Only


Formula: C₈H₆O₆
Melting Pt: 309…310 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02181105
CAS Number: 19829-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty