Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Triaminopyrimidine 97%

Supplier: AMBEED, INC
Warning

A256073-5G A256073-25G A256073-100G A256073-10G A256073-5G
103809-816EA 175 USD InStock
103809-816 103852-098 77091-574 77289-132 77289-134 77304-670
2,4,6-Triaminopyrimidine 97%
2,4,6-Triaminopyrimidine
Precaución: for Research Use Only


Formula: C₄H₇N₅
Melting Pt: 249…251 °C
Flash Pt: 308.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00006100
CAS Number: 1004-38-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty