Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromoterephthalic acid 98%

Supplier: AMBEED, INC
Warning

A131941-1G A131941-100G A131941-10G A131941-500G A131941-1G A131941-25G A131941-5G
103801-658EA 5 USD InStock
103801-658 77190-084 77190-086 77190-088 77298-520 77314-190 77314-192
2,5-Dibromoterephthalic acid 98%
2,5-Dibromoterephthalic acid
Precaución: for Research Use Only


Formula: C₈H₄Br₂O₄
Melting Pt: >320 °C
Flash Pt: 222.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00204593
CAS Number: 13731-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty