Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-1,8-naphthyridine 95%

Supplier: AMBEED, INC
Warning

A141393-1G A141393-100MG A141393-250MG A141393-1G A141393-5G
103802-990EA 246 USD InStock
103802-990 76823-458 76823-460 77316-146 77316-148
2,5-Dichloro-1,8-naphthyridine 95%
2,5-Dichloro-1,8-naphthyridine
Precaución: for Research Use Only


Formula: C₈H₄Cl₂N₂
Boiling Pt: 300.4 °C (760 mm Hg)
Flash Pt: 163.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13181698
CAS Number: 91870-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty