Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dioxopyrrolidin-1-yl ((3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl) carbonate 95%

Supplier: AMBEED, INC
Warning

A222665-1G A222665-5G A222665-100MG A222665-1G
103818-990EA 19 USD InStock
103818-990 103849-868 76826-228 76826-230 77303-628
2,5-Dioxopyrrolidin-1-yl ((3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl) carbonate 95%
2,5-Dioxopyrrolidin-1-yl ((3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl) carbonate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₇
Boiling Pt: 391.8 °C (760 mm Hg)
Flash Pt: 190.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031467
CAS Number: 253265-97-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty