Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,7-Diazaspiro[4.5]decan-1-one hydrochloride 97%

Supplier: AMBEED, INC
Warning

A401557-1G A401557-5G A401557-100MG A401557-250MG
103827-058EA 110 USD InStock
103827-058 103838-096 77033-662 77033-664
2,7-Diazaspiro[4.5]decan-1-one hydrochloride 97%
2,7-Diazaspiro[4.5]decan-1-one hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD08460858
CAS Number: 1187173-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty